Ac4ManNAl ≥95%

Supplier: AFG Bioscience

Synonyms: N-(4-pentynoyl)-mannosamine tetraacetate

C52021-100MG C52021-50MG C52021-25MG
76619-776EA 550.84 USD
76619-776 76619-774 77888-843
Ac4ManNAl ≥95%
Ac4ManNAl
Formula: C₁₉H₂₅NO₁₀
MW: 427.4 g/mol
Storage Temperature: Freezer
CAS Number: 935658-93-8

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR