α-Man-PEG3-Alkyne 95%

Supplier: AFG Bioscience

Synonyms: 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl α-D-Mannopyranoside

C52005-50MG C52005-25MG C52005-100MG C52005-10MG
76619-734EA 492.92 USD
76619-734 76619-732 77888-831 77888-832
α-Man-PEG3-Alkyne 95%
α-Man-PEG3-Alkyne
Formula: C₁₅H₂₆O₉
Storage Temperature: Freezer
CAS Number: 1442747-33-2

Order Now

Specification Test Results

Purity Limit 95%
Appearance White powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR