2-Azidoethyl LacNAc ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-Azidoethyl N-acetyl-D-lactosamine

C51783-25MG C51783-10MG
76619-694EA 1490.53 USD
76619-694 76619-692
2-Azidoethyl LacNAc ≥95%
2-Azidoethyl LacNAc
Formula: C₁₆H₂₈N₄O₁₁
MW: 452.41 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19981037
CAS Number: 338971-38-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR