2-Azido-2-deoxy-D-galactose 95%

Supplier: AFG BIOSCIENCE LLC

C51521-1G C51521-500MG
76619-476EA 999.96 USD
76619-476 76619-474
2-Azido-2-deoxy-D-galactose 95%
2-Azido-2-deoxy-D-galactose
Formula: C₆H₁₁N₃O₅
MW: 205.17 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09750682
CAS Number: 68733-26-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR