Fmoc-L-Thr (Gal β(1-3)GalNAc)-OH, peracetate ≥95%

Supplier: AFG Bioscience

C51032-100MG C51032-250MG C51032-25MG
76619-352EA 2253.43 USD
76619-352 76619-354 76619-350
Fmoc-L-Thr (Gal β(1-3)GalNAc)-OH, peracetate ≥95%
Fmoc-L-Thr (Gal β(1-3)GalNAc)-OH, peracetate
Formula: C₄₅H₅₄N₂O₂₁
Storage Temperature: Freezer
MDL Number: MFCD00167470
CAS Number: 125760-33-0

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR