2α-Mannobiose ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 2-O-α-D-Mannopyranosyl-D-mannopyranose

C50360-100MG C50360-250MG C50360-50MG
76619-102EA 500.76 USD
76619-102 76619-104 76619-100
2α-Mannobiose ≥95%
2α-Mannobiose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01310887
CAS Number: 15548-39-7

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White Powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR