1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate 97%

Supplier: AFG Bioscience

C51537-1G C51537-2G C51537-5G
76619-538EA 124.8 USD
76619-538 76619-540 76619-542
1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate 97%
1-Azido-1-deoxy-β-D-glucopyranoside tetraacetate
Formula: C₁₄H₁₉N₃O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00216966
CAS Number: 13992-25-1

Order Now

Specification Test Results

Purity Limit 97%
Appearance White Crystalline Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR