6-Azido-6-deoxy-D-glucopyranose 95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: 6-Azido-6-deoxy-D-glucose

C51533-100MG C51533-250MG
76619-524EA 250.38 USD
76619-524 76619-526
6-Azido-6-deoxy-D-glucopyranose 95%
6-Azido-6-deoxy-D-glucopyranose
Formula: C₆H₁₁N₃O₅
MW: 205.169 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03265537
CAS Number: 20847-05-6

Order Now

Specification Test Results

Purity Limit 95%
Appearance light-yellow sticky oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR