N-Glycolylneuraminic acid ≥95%

Supplier: AFG BIOSCIENCE LLC

Synonyms: Neu5Gc

C53623-100MG C53623-25MG
76619-944EA 500.76 USD
76619-944 76619-942
N-Glycolylneuraminic acid ≥95%
N-Glycolylneuraminic acid
Formula: C₁₁H₁₉NO₁₀
MW: 325.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00057551
CAS Number: 1113-83-3

Order Now

Specification Test Results

Purity Limit > 95%
Appearance White To Off-White Solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR