3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥95%
Supplier: AFG BIOSCIENCE LLC
C50720-1G
C50720-5G
76619-206EA
445.98
USD
76619-206
76619-208
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose ≥95%
3,4,6-Tri-O-benzyl-2-deoxy-D-glucopyranose
Formula:
C₂₇H₃₀O₅ MW: 434.53 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06797170 CAS Number: 132732-60-6 |
Specification Test Results
Purity Limit | > 95% |
Appearance | Powder or Crystals |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...