2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide 97%
Supplier: AFG BIOSCIENCE LLC
Synonyms:
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl Azide, 2-Acetamido-2-deoxy-β-D-glucopyranosyl azide 3,4,6-triacetate
C51570-1G
C51570-2G
C51570-5G
76619-594EA
120.18
USD
76619-594
76619-596
76619-598
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl azide 97%
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl azide
Formula:
C₁₄H₂₀N₄O₈ MW: 372.33 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00216968 CAS Number: 6205-69-2 |
Specification Test Results
Purity Limit | 97% |
Appearance | White Solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...