MC-Val(D8)-Cit-PAB-OH ≥95%
Supplier: AFG Bioscience
C56092-100MG
C56092-25MG
76620-252EA
1232.31
USD
76620-252
76620-250
MC-Val(D8)-Cit-PAB-OH ≥95%
MC-Val(D8)-Cit-PAB-OH
Specification Test Results
Purity Limit | >95% |
Appearance | Off-white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...