FrameStar® PCR Plates, 96-Well, Semi-Skirted, Roche Style, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0951 4TI-0950/C 4TI-0952 4TI-0953 4TI-0954 4TI-0954/0560
76619-080CS 346.62 USD
76619-080 76629-138 76629-136 76628-984 76628-992 76629-034
FrameStar® PCR Plates, 96-Well, Semi-Skirted, Roche Style, Azenta Life Sciences
Microplates PCR Plates
Low profile, 0.1 ml polypropylene wells, polycarbonate frame, cut corner H12, working volume: <100 µl, total well capacity: 200 µl. Ideal for use with robotic systems. Compatible with Roche LightCycler 96 and 480 (with 96-well block). Compatible with standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
The FrameStar Roche Style plates are designed to achieve optimized assay conditions on the Roche LightCycler® 96 and 480 (with 96 well block).
This particular style of plate is in a low profile 96-well format, perfect for reaction volumes of 10-100 µl. The wells are shorter than “standard” profile wells, which decrease the “dead space” between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction. This is especially recommended for reaction volumes below 20 µl.
The semi-skirted frame allows for labeling or barcoding.
Información para pedidos: Available barcoded upon request.
Envase: 50 plates per case.