FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® FastPlate Style, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0910/C 4TI-0912 4TI-0912/10P 4TI-0910/C/10P 4TI-0911
76619-078CS 307.46 USD
76619-078 76619-074 76628-996 76629-160 76628-986
FrameStar® PCR Plates, 96-Well, Semi-Skirted, ABI® FastPlate Style, Azenta Life Sciences
Microplates PCR Plates
Low profile, 0.1 ml polypropylene wells, polycarbonate frame with upstand, cut corner A1, working volume: <100 µl, total well capacity: 200 µl. Ideal for use with robotic systems. Compatible with ABI® Fast block thermal cyclers. Compatible with standard multichannel pipettes.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
This semi-skirted low profile plate is recommended for use with ABI® Fast block thermal cyclers.
The low profile wells of this plate are shorter than “standard” profile wells, which decrease the “dead space” between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction. Especially recommended for reaction volumes below 20 µl.
The rigid polycarbonate skirt of this design eliminates warping and distortion during PCR, and makes it ideal for use with robotic systems. Its skirt also allows for labeling or barcoding.
Información para pedidos: Available barcoded upon request.
Envase: 50 plates per case.