FrameStar® PCR Plates, 384-Well, Fully Skirted, Azenta Life Sciences
Supplier: AZENTA US, INC
4TI-0384/C 4TI-0386 4TI-0387 4TI-0384 4TI-0384/B 4TI-0384/G 4TI-0384/R 4TI-0384/X 4TI-0385 4TI-LB0384/C
76619-072CS 307.46 USD
76619-072 76629-078 76629-048 76629-018 76629-142 76629-066 76628-980 76629-014 76629-028 77321-778
FrameStar® PCR Plates, 384-Well, Fully Skirted, Azenta Life Sciences
Microplates PCR Plates
Polypropylene wells, polycarbonate frame, cut corner A24; working volume: <30 µl, total well capacity: 55 µl. Ideal for use with robotic systems. Compatible with standard multichannel pipettes and compatible with the majority of 384 well block PCR, qPCR and sequencing instruments.
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Stackable
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Rigid polycarbonate frame for added mechanical stability
  • Significantly reduced thermal expansion and sample evaporation for Improved consistency in PCR results
  • Improved seal integrity due to thermal stability of frame
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
The FrameStar 384-Well Skirted PCR Plates are designed for high-throughput PCR and are compatible with the majority of 384 well block PCR, qPCR and sequencing instruments.
Rigid 2-component design eliminates warping and distortion during PCR making it ideal for use with robotic systems and the skirt allows for labeling or barcoding.
Información para pedidos: Available barcoded upon request.
Envase: 50 plates per case.