3-Fluoro-5-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Fluoro-5-hydroxybenzotrifluoride

PC4376P-1G PC4376P-25G PC4376P-5G
77415-366EA 41.78 USD
77415-366 77415-368 77415-370
3-Fluoro-5-(trifluoromethyl)phenol 98%
3-Fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061288
CAS Number: 172333-87-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR