2-Chloro-3-hydrazinopyrazine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-3-hydrazinylpyrazine

OR61106-250MG OR61106-5G
77410-698EA 20.28 USD
77410-698 77410-700
2-Chloro-3-hydrazinopyrazine
2-Chloro-3-hydrazinopyrazine
Formula: C₄H₈Cl₂N₄
MW: 144.5600 g/mol
Melting Pt: 154…159 °C
Storage Temperature: Ambient
MDL Number: MFCD08272804
CAS Number: 63286-28-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR