MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 2-hydroxy-3-morpholinopropane-1-sulphonate
462141000
462145000
TS462141000EA
213
USD
TS462141000
TS462145000
MOPSO sodium salt (3-(4-morpholino)-2-hydroxypropanesulfonic acid sodium salt)
MOPSO sodium salt (3-(4-Morpholino)-2-hydroxypropanesulphonic acid sodium salt)
- Bioactive
Formula:
C₇H₁₄NNaO₅S MW: 247.2445 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070007 CAS Number: 79803-73-9 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Total base | ≥98.5 % |
Water | ≤0.5 % |
UV | at 290 nm A: ≤0.1 (33% W/W) |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...