1-(Mercaptomethyl)cyclopropaneacetic acid 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: [1-(Thiomethyl)cycloprop-1-yl]acetic acid, 2-(1-(Mercaptomethyl)cyclopropyl)acetic acid

OR3739-25G OR3739-5G
77409-358EA 117.16 USD
77409-358 77409-360
1-(Mercaptomethyl)cyclopropaneacetic acid 97%
1-(Mercaptomethyl)cyclopropaneacetic acid
Formula: C₆H₁₀O₂S
MW: 146.20 g/mol
Melting Pt: 42…46 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03411600
CAS Number: 162515-68-6
EINECS: 420-240-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR