Ethyl cyanoacetate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR28668-100G OR28668-25G
77408-154EA 117.16 USD
77408-154 77408-156
Ethyl cyanoacetate 98%
Ethyl cyanoacetate
Formula: C₅H₇NO₂
Boiling Pt: 208…210 °C
Melting Pt: –23 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
UN: 3334
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR