2-Methoxyphenyl isocyanate 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Isocyanato-2-methoxybenzene

OR0583-250MG OR0583-5G
77402-152EA 20.26 USD
77402-152 77402-154
2-Methoxyphenyl isocyanate 95%
2-Methoxyphenyl isocyanate

  • May form small amount of sediment.

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 94…96 °C/10mmHg
Density: 1.122 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002004
CAS Number: 700-87-8
EINECS: 211-853-5
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR