3-Nitro-o-cresol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Methyl-3-nitrophenol, 1-Hydroxy-2-methyl-3-nitrobenzene

OR27719-1G OR27719-5G
77407-964EA 20.26 USD
77407-964 77407-966
3-Nitro-o-cresol 98%
3-Nitro-o-cresol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: 146…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00007241
CAS Number: 5460-31-1
EINECS: 226-739-0
UN: 2446
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR