4'-Isopropylacetophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: p-Isopropylacetophenone, 4-Isopropylacetophenone

OR2867-100G OR2867-25G OR2867-5G
77408-160EA 206.82 USD
77408-160 77408-162 77408-164
4'-Isopropylacetophenone 98%
4'-Isopropylacetophenone
Formula: C₁₁H₁₄O
MW: 162.23 g/mol
Boiling Pt: 238 °C
Density: 0.97 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00048297
CAS Number: 645-13-6
EINECS: 211-433-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR