5-Chloro-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Chloroindole-3-carboxaldehyde

OR28433-100MG OR28433-5G
77408-080EA 20.26 USD
77408-080 77408-082
5-Chloro-1H-indole-3-carbaldehyde
5-Chloro-1H-indole-3-carbaldehyde
Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 211…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00175291
CAS Number: 827-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR