1,3-Benzoxazol-5-amine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-1,3-benzoxazole5-Aminobenzoxazole

OR15063-1G OR15063-5G OR15063-250MG
77405-268EA 26 USD
77405-268 76567-438 76567-436
1,3-Benzoxazol-5-amine
1,3-Benzoxazol-5-amine
Formula: C₇H₆N₂O
MW: 134.135 g/mol
Melting Pt: 64…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05256201
CAS Number: 63837-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR