2-(4-Formylphenyl)isonicotinonitrile 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR14211-500MG
OR14211-5G
77404-854EA
241.46
USD
77404-854
77404-856
2-(4-Formylphenyl)isonicotinonitrile 95%
2-(4-Formylphenyl)isonicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula:
C₁₃H₈N₂O Melting Pt: 175…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200959 CAS Number: 253801-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...