4-tert-Butyl-3-methoxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR13716-500MG OR13716-5G
77404-722EA 34.64 USD
77404-722 77404-724
4-tert-Butyl-3-methoxybenzoic acid
4-tert-Butyl-3-methoxybenzoic acid
Formula: C₁₂H₁₆O₃
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD06656116
CAS Number: 79822-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR