2-Chloro-1,3-benzothiazole-6-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-6-benzothiazolecarboxylic acid

OR12055-10G OR12055-5G
77404-136EA 206.82 USD
77404-136 77404-138
2-Chloro-1,3-benzothiazole-6-carboxylic acid
2-Chloro-1,3-benzothiazole-6-carboxylic acid
Formula: C₈H₄ClNO₂S
MW: 213.64 g/mol
Melting Pt: 220…221 °C
Storage Temperature: Ambient
MDL Number: MFCD08689736
CAS Number: 3855-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR