Evergreen 48-Well Deep Well Plates, Caplugs Evergreen
Supplier: Caplugs
222-8672-08C 290-8560-08C
76521-416CT 140.55 USD
76521-416 76521-418
Evergreen 48-Well Deep Well Plates, Caplugs Evergreen
Microplates Deep Well Microplates
Caplugs evergreen 48-well deep well plates have the same footprint as the regular microplates and are available in sterile and non sterile versions.
  • Suitable for dilution and storage
  • Distinct alpha-numeric well designations
  • Well volume: 5 ml
  • Each well bottom has rounded corners and chamfered edges to eliminate dead space during mixing
  • Polypropylene lids are available
  • EZ Seal™ film compatible
24 deep-well and 96 deep-well units are also available.