N4-Benzoylcytosine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

46054-0050 46054-0250
TS46054-0050EA 37 USD
TS46054-0050 TS46054-0250
N4-Benzoylcytosine 98%
N4-Benzoylcytosine
Formula: C₁₁H₉N₃O₂
MW: 215.21 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00239434
CAS Number: 26661-13-2

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥19.04 to ≤20.01 % Nitrogen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR