Hexyl nicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nicotinic Acid Hexyl Ester

46009-0050 46009-0250
TS46009-0050EA 36 USD
TS46009-0050 TS46009-0250
Hexyl nicotinate 98%
Hexyl nicotinate
Formula: C₁₂H₁₇NO₂
MW: 207.27 g/mol
Boiling Pt: 329 °C (760 mmHg)
Density: 1.02 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00023587
CAS Number: 23597-82-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR