2-Chloro-3-methoxybenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Chloro-m-anisic acid
45990-0010
45990-0050
TS45990-0010EA
72
USD
TS45990-0010
TS45990-0050
2-Chloro-3-methoxybenzoic acid 97%
2-Chloro-3-methoxybenzoic acid
Formula:
C₈H₇ClO₃ MW: 186.59 g/mol Melting Pt: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD07784399 CAS Number: 33234-36-5 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder or crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 160 to 165 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...