2-Methylpyrimidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
45986-0010
45986-0050
TS45986-0010EA
98
USD
TS45986-0010
TS45986-0050
2-Methylpyrimidine 98%
2-Methylpyrimidine
Formula:
C₅H₆N₂ Boiling Pt: 130…132 °C (760 mmHg) Melting Pt: –6 °C Density: 1.0132 g/cm³ Flash Pt: 37.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00128182 CAS Number: 5053-43-0 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4890 to 1.4940 (20°C, 589 nm) |
Water | ≤2.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...