4,6-Diaminopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,6-Pyrimidinediamine

45980-0010 45980-0050
TS45980-0010EA 41 USD
TS45980-0010 TS45980-0050
4,6-Diaminopyrimidine 98%
4,6-Diaminopyrimidine
Formula: C₄H₆N₄
MW: 110.12 g/mol
Melting Pt: 271 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00044572
CAS Number: 2434-56-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR