2-Chloro-5-nitrophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

45904-0050 45904-0250
TS45904-0050EA 66 USD
TS45904-0050 TS45904-0250
2-Chloro-5-nitrophenol 98%
2-Chloro-5-nitrophenol
Formula: C₆H₄ClNO₃
MW: 173.56 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD01571825
CAS Number: 619-10-3

Order Now

Specification Test Results

Appearance Off-white to brown Powder or solid
Infrared spectrum Conforms
Melting point 117 to 123 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR