4-Azaphthalide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Hydroxymethyl)nicotinic acid gamma-lactone, Furo[3,4-b]pyridin-5(7H)-one

45845-0010 458450050 458450250
TS45845-0010EA 111 USD
TS45845-0010 TS458450050 TS458450250
4-Azaphthalide 98%
4-Azaphthalide
Formula: C₇H₅NO₂
MW: 135.12 g/mol
Melting Pt: 143…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860224
CAS Number: 5657-51-2

Order Now

Specification Test Results

Appearance White to light brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 140 to 146 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR