Isothiazole 97%

Supplier: Thermo Scientific Chemicals
Danger

45842-0010 458420050
TS45842-0010EA 191 USD
TS45842-0010 TS458420050
Isothiazole 97%
Isothiazole
Formula: C₃H₃NS
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00020818
CAS Number: 288-16-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR