2-Bromo-1-chloro-4-iodobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45838-0010
45838-0050
TS45838-0010EA
101
USD
TS45838-0010
TS45838-0050
2-Bromo-1-chloro-4-iodobenzene 98%
2-Bromo-1-chloro-4-iodobenzene
Formula:
C₆H₃BrCl Melting Pt: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD07780667 CAS Number: 31928-46-8 |
Specification Test Results
Appearance | Yellow to brown Powder or crystalline powder or low melting solid |
Infrared spectrum | Conforms |
Melting point | 39 to 44 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...