4-Amino-3-fluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Fluoro-4-hydroxyaniline

45831-0010 45831-0050
TS45831-0010EA 97 USD
TS45831-0010 TS45831-0050
4-Amino-3-fluorophenol 98%
4-Amino-3-fluorophenol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 137…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077452
CAS Number: 399-95-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Appearance Light yellow to brown Powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR