4-Nitro-o-toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

45781-0050 45781-0250 45781-1000
TS45781-0050EA 72 USD
TS45781-0050 TS45781-0250 TS45781-1000
4-Nitro-o-toluic acid 98%
4-Nitro-o-toluic acid
Formula: C₈H₇NO₄
Melting Pt: 151…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00210697
CAS Number: 1975-51-5

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR