2,3,6,7,10,11-Hexahydroxytriphenylene 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triphenylene-2,3,6,7,10,11-hexaol

45701-0010 45701-0050
TS45701-0010EA 121 USD
TS45701-0010 TS45701-0050
2,3,6,7,10,11-Hexahydroxytriphenylene 95%
2,3,6,7,10,11-Hexahydroxytriphenylene
Formula: C₁₈H₁₂O₆
MW: 324.29 g/mol
Melting Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD01321170
CAS Number: 4877-80-9

Order Now

Specification Test Results

Appearance Grey to purple-grey Powder
Infrared spectrum Conforms
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR