1,10-Phenanthroline-5,6-dione 98%

Supplier: Thermo Scientific Chemicals
Warning

45608-0010 45608-0050
TS45608-0010EA 155 USD
TS45608-0010 TS45608-0050
1,10-Phenanthroline-5,6-dione 98%
1,10-Phenanthroline-5,6-dione
Formula: C₁₂H₆N₂O₂
Melting Pt: 257…258 °C
Storage Temperature: Ambient
MDL Number: MFCD00014473
CAS Number: 27318-90-7

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 255 to 260 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR