2-Pyrimidinecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine

45583-0010 45583-0050
TS45583-0010EA 63 USD
TS45583-0010 TS45583-0050
2-Pyrimidinecarboxylic acid 95%
2-Pyrimidinecarboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.1 g/mol
Melting Pt: 190…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00856161
CAS Number: 31519-62-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR