4-(Hydroxymethyl)benzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyanobenzyl alcohol

45465-0010 45465-0050 45465-0250
TS45465-0010EA 68 USD
TS45465-0010 TS45465-0050 TS45465-0250
4-(Hydroxymethyl)benzonitrile 97%
4-(Hydroxymethyl)benzonitrile
Formula: C₈H₇NO
MW: 133.15 g/mol
Melting Pt: 41…42 °C
Storage Temperature: Ambient
MDL Number: MFCD00870633
CAS Number: 874-89-5
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to white to yellow Low melting solid
Infrared spectrum Conforms
Melting point 38 to 44 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR