5-Thiazolecarboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

45454-0010 45454-0050
TS45454-0010EA 158 USD
TS45454-0010 TS45454-0050
5-Thiazolecarboxylic acid 98%
5-Thiazolecarboxylic acid
Formula: C₄H₃NO₂S
MW: 129.1371 g/mol
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD03428539
CAS Number: 14527-41-4

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR