6-Chloro-m-anisidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole

45378-0050 45378-0250
TS45378-0050EA 68 USD
TS45378-0050 TS45378-0250
6-Chloro-m-anisidine 98%
6-Chloro-m-anisidine
Formula: C₇H₈ClNO
MW: 157.6 g/mol
Boiling Pt: 135…137 °C (12 mmHg)
Melting Pt: 23…27 °C
Density: 1.261 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00047830
CAS Number: 2401-24-3

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Low melting solid or liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR