2,4-Dibromopyridine 97%

Supplier: Thermo Scientific Chemicals
Danger

45370-0010 45370-0050
TS45370-0010EA 63 USD
TS45370-0010 TS45370-0050
2,4-Dibromopyridine 97%
2,4-Dibromopyridine
Formula: C₅H₃Br₂N
Melting Pt: 36…38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01859720
CAS Number: 58530-53-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Off-white to yellow to brown Low melting solid
Infrared spectrum Conforms
Melting point 35 to 40 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR