2-(Chloromethyl)benzoxazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Chloromethyl)benzo[d]oxazole, 2-(Chloromethyl)-1,3-benzoxazole

45338-0010 45338-0050
TS45338-0010EA 144 USD
TS45338-0010 TS45338-0050
2-(Chloromethyl)benzoxazole 97%
2-(Chloromethyl)benzoxazole
Formula: C₈H₆ClNO
MW: 167.5900 g/mol
Boiling Pt: 80 °C (.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD05664964
CAS Number: 41014-43-1

Order Now

Specification Test Results

Appearance Colorless to orange Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR