5-Amino-1-methyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

45261-0250 45261-1000
TS45261-0250EA 238 USD
TS45261-0250 TS45261-1000
5-Amino-1-methyl-1H-pyrazole 97%
5-Amino-1-methyl-1H-pyrazole
Formula: C₄H₇N₃
MW: 97.12 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00068156
CAS Number: 1192-21-8

Order Now

Specification Test Results

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 69 to 77 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR