Imidazo[1,2-a]pyridine-6-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45174-0010
45174-0100
TS45174-0010EA
60
USD
TS45174-0010
TS45174-0100
Imidazo[1,2-a]pyridine-6-carboxylic acid 97%
Imidazo[1,2-a]pyridine-6-carboxylic acid
Formula:
C₈H₆N₂O₂ Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD07021498 CAS Number: 139022-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...